Anti-GUCA1C Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-13308R-CY7
BOSSBS-13308R-CY7EA 900 EUR InStock
BOSSBS-13308R-CY7
Anti-GUCA1C Rabbit Polyclonal Antibody (Cy7®)
Anticorps
The intracellular stimulation of guanylate cyclase (GC) by calcium, a key event in the recovery of the dark state of rod photoreceptors after exposure to light, is mediated by guanylate cyclase-activating proteins (GCAP). GCAPs are calcium-binding proteins belonging to the calmodulin superfamily and are specifically expressed in retina. GCAP3 (Guanylyl cyclase-activating protein 3), also known as GUCA1C (Guanylate cyclase activator 1C), is a 209 amino acid EF-hand calcium binding protein that is activated by the decrease in calcium from the absorption of light by rhodopsin. Activation ofMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité