Anti-GLI4 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-13373R-CY3
BOSSBS-13373R-CY3EA 900 EUR InStock
BOSSBS-13373R-CY3
Anti-GLI4 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. GLI-4, also known as HKR4, is a 376 amino acid protein that localizes to the nucleus and contains seven C2H2-type zinc fingers. Belonging to the krueppel C2H2-type zinc-finger protein family, GLI-4 may function as a transcriptional regulator,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité