Anti-GSTK1 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-13399R-CY7
BOSSBS-13399R-CY7EA 900 EUR InStock
BOSSBS-13399R-CY7
Anti-GSTK1 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Members of the glutathione S-transferase (GST) family of proteins function in the detoxification of xenobiotics to protect cells against toxicant-induced damage. There are eight families of GST proteins, namely alpha, zeta, theta, kappa, mu, pi, sigma and omega, each of which are composed of proteins that have a variety of functions throughout the cell. GSTK1 (glutathione S-transferase kappa 1), also known as glutathione S-transferase subunit 13 (GST 13-13) or GSTK1-1, is a 226 amino acid ubiquitously expressed protein belonging to the kappa class of the GST superfamily that functions inMore Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-13399R-CY7
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité