Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-13447R-A680
BOSSBS-13447R-A680EA 890 EUR InStock
BOSSBS-13447R-A680
Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Glycogen synthesis is initiated by the autoglucosylation of Glycogenin-1. Specifically, Glycogenin-1 glucosylates itself to begin the synthesis of glycogen in mammalian skeletal muscle. It acts as the primer to which further glucose monomers may be added. All of the Glycogenin-1 molecules contain at least one glucosyl residue before autoglucosylation begins. The first step of the glycogen synthesis occurs when a glucose molecule from UDP-glucose binds to the hydroxyl group of Tyr 194 on the Glycogenin-1 molecule. Using its glucosyltransferase activity, Glycogenin-1 adds more glucoses, each one coming from UDP-glucose. The glycosylation process reaches a plateau when five new glucose residues have been added, at which point glycogen synthase (GS) takes over and further elongates the chain. Glycogenin-1 remains covalently attached to the reducing end of the glycogen molecule.
Type: Primary
Antigen: Glycogenin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/350
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité