Anti-GMEB2 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-13456R-CY5
BOSSBS-13456R-CY5EA 900 EUR InStock
BOSSBS-13456R-CY5
Anti-GMEB2 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
GMEB-2 is a DNA-binding protein that plays a crucial role modulating transcription upon activation by steroid hormones. GMEB-2 is ubiquitously expressed with preferential expression in developmentally important tissues, such as testis, bone marrow, placenta and fetal tissues. It localizes to the nucleus and cytoplasm and contains a SAND domain near its N-terminus and a C-terminal coiled coil structure. GMEB-2 functions as a homodimer or as a heterodimer with GMEB-1. The formed complex specifically binds to glucocorticoid modulatory elements (GME) in the promoter region of target genes andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité