Anti-GNGT1 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-13470R-CY3
BOSSBS-13470R-CY3EA 900 EUR InStock
BOSSBS-13470R-CY3
Anti-GNGT1 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Heterotrimeric guanine nucleotide-binding proteins (G proteins) transduce extracellular signals received by transmembrane receptors to effector proteins. Transducin is a guanine nucleotide-binding protein found specifically in rod outer segments, where it mediates activation by rhodopsin of a cyclic GTP-specific (guanosine monophosphate) phosphodiesterase. Transducin is also referred to as GMPase. GNGT1 encodes the gamma subunit of transducin (Hurley et al., 1984 [PubMed 6438626]; Scherer et al., 1996 [PubMed 8661128]).[supplied by OMIM, Mar 2008].
Recommended Dilutions: IF(IHC-P): 1:50-20More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité