Anti-FADD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-1349R-A647
BOSSBS-1349R-A647EA 921 EUR InStock
BOSSBS-1349R-A647
Anti-FADD Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
FADD (Fas Associated Death Domain) is an apoptosis adapter molecule enabling transduction of the apoptosis signal initiated via the FasL/Fas receptor interaction. The protein contains a C terminal death domain that interacts with the Fas receptor death domain. The N terminus contains a death effectors domain (DED) which recruits caspase to the death inducing signaling complex (DISC) and initiates the apoptotic caspase cascade. Recruitment of Caspase 8 to the Fas receptor results in oligomerization of the Caspase 8 protein, which in turn drives its autoactivation through self-cleavage.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité