Anti-GPR84 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-13507R-A647
BOSSBS-13507R-A647EA 893 EUR InStock
BOSSBS-13507R-A647
Anti-GPR84 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 84 (GPR84), a member of the GCPR 1 family, is an orphan GCPR expressed in bone marrow, brain, heart, muscle,More Product Information
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-13507R-A647
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité