Anti-GPR20 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-13523R-CY3
BOSSBS-13523R-CY3EA 900 EUR InStock
BOSSBS-13523R-CY3
Anti-GPR20 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. G protein-coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR20 is a 358 amino acidMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité