Anti-Guanylyl Cyclase alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-13544R-A750
BOSSBS-13544R-A750EA 890 EUR InStock
BOSSBS-13544R-A750
Anti-Guanylyl Cyclase alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase. The soluble forms, known as GCS or sGC, act as receptors for nitric oxide. The membrane-bound receptor forms, known as GC, are peptide hormone receptors. GCS, a cGMP-synthesizing enzyme, is the major receptor for the neurotransmitter nitric oxide (NO). It plays a crucial role in smooth muscle contractility, platelet reactivity and neurotransmission. GCS is a heme containing heterodimer, consisting of one alpha subunit, designated GCS-alpha-1, and one beta subunit. TheMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité