Anti-ZBED5 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-13555R-CY7
BOSSBS-13555R-CY7EA 900 EUR InStock
BOSSBS-13555R-CY7
Anti-ZBED5 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
ZBED5 contains 1 BED type zinc finger and it may be derived from an ancient transposon that has lost its ability to translocate. This gene is unusual in that its coding sequence is mostly derived from Charlie like DNA transposon. There is mRNA and EST evidence to suggest that this gene is transcribed. The encoded protein shares 70% identity with Charlie 1 transposase, however, this gene does not appear to be an active DNA transposon as it is not flanked by terminal inverted repeats. The exact function of this gene product is not known.
Recommended Dilutions: IF(IHC-P): 1:50-200
Collapse All
Cdt Numéro de catalogue VWR Quantité
100 µl BOSSBS-13555R-CY7
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité