Anti-IFI30 Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-13591R-CY3
BOSSBS-13591R-CY3EA 900 EUR InStock
BOSSBS-13591R-CY3
Anti-IFI30 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Proteins internalized into the endocytic pathway are usually degraded. Efficient proteolysis requires denaturation, induced by acidic conditions within lysosomes, and reduction of inter- and intrachain disulfide bonds. Cytosolic reduction is mediated enzymatically by thioredoxin. In the endocytic pathway, reduction of protein disulfide bonds is important for the generation of MHC class II-peptide complexes. This process is catalyzed by a gamma-interferon-inducible thiol reductase (GILT). GILT is synthesized as a precursor, and following delivery to MHC class II-containing compartments (MIICs)More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité