Anti-DPPA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Fournisseur: Bioss
BS-13595R-A647
BOSSBS-13595R-A647EA 893 EUR InStock
BOSSBS-13595R-A647
Anti-DPPA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorps
DPPA4 (developmental pluripotency associated 4) is a 304 amino acid protein that localizes to the nucleus and is thought to be involved in the maintenance of cell pluripotentiality during embryogenesis. The gene encoding DPPA4 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, vonMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité