Anti-ZNF323 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-13604R-HRP
BOSSBS-13604R-HRPEA 900 EUR InStock
BOSSBS-13604R-HRP
Anti-ZNF323 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. As a member of the Krüppel C2H2-type zinc-finger protein family, ZNF323 (zinc finger protein 323) is a 406 amino acid protein containing six C2H2-type zinc fingers and one SCAN box domain. Specifically, C2H2-type zinc fingers function to bind DNA, while SCAN box domains are thought to participate in protein-protein interactions. Therefore, it is probable that ZNF323 functions as a transcription factor. With highest expressionMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité