Anti-BAIAP2L2 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-13684R
BOSSBS-13684REA 725 EUR InStock
BOSSBS-13684R
Anti-BAIAP2L2 Rabbit Polyclonal Antibody
Anticorps
The Src homology 3 (SH3) domain is a highly conserved 60 amino acid protein domain that is organized into a beta-barrel fold consisting of five or six beta strands arranged as two tightly packed anti-parallel beta sheets. This domain is found in proteins that mediate assembly of specific protein complexes and interact with other proteins, specifically recognizing proline-rich regions. BAIAP2L2 (Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 2) is a 529 amino acid protein containing an SH3 domain, suggesting that it may function as an adaptor protein. BAIAP2L2 alsoMore Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 μg 100 µl BOSSBS-13684R
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité