Anti-MPP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-13689R-A350
BOSSBS-13689R-A350EA 893 EUR InStock
BOSSBS-13689R-A350
Anti-MPP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
MPP2 (Palmitoylated membrane protein 2) is a member of a family of membrane associated proteins termed MAGUKs (membrane associated guanylate kinase homologs). These proteins interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. MPP2 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MPP2
Clonality: Polyclonal
Clone:More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité