Anti-NOSTRIN Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-13694R-A680
BOSSBS-13694R-A680EA 890 EUR InStock
BOSSBS-13694R-A680
Anti-NOSTRIN Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
NOSTRIN (nitric oxide synthase trafficker isoform 1), also known as endothelial nitric oxide synthase traffic inducer, is a member of the Pombe Cdc15 homology (PCH) family of proteins. NOSTRIN is expressed in the vascular endothelial cells of highly vascularized tissues such as placenta, lung, kidney and heart. It consists of an N-terminal Cdc15 domain with an FCH (Fes/CIP homology) region, two coiled coil domains and a C-terminal SH3 domain. NOSTRIN typically exists as a trimer. It functions as an adaptor protein binding to caveolin-1 via an internal domain and NOS3 via its SH3 domain,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité