Anti-PAG1 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-13695R
BOSSBS-13695REA 725 EUR InStock
BOSSBS-13695R
Anti-PAG1 Rabbit Polyclonal Antibody
Anticorps
The Src family of protein tyrosine kinases (Src-PTKs) is important in the regulation of growth and differentiation of eukaryotic cells. The activity of Src-PTKs in cells of different types is negatively controlled by Csk. Csk binding protein (Cbp), also designated phosphoprotein associated with glycosphingo-lipid-enriched microdomains (GEMs) or PAG, is a ubiquitously expressed transmembrane phosphoprotein that binds specifically to the SH2 domain of Csk. Cbp is involved in the membrane localization of Csk and in Csk-mediated inhibition of c-Src. In the plasma membrane, Cbp is exclusivelyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité