Anti-RAGEF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-13698R-A488
BOSSBS-13698R-A488EA 893 EUR InStock
BOSSBS-13698R-A488
Anti-RAGEF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
RAPGEF6 is a guanine nucleotide exchange factor (GEF) that is expressed in a variety of tissues. Localizing to the cytoplasm and translocated to the plasma membrane upon ligand binding, RAPGEF6 contains an N-terminal Ras-GEF domain, a cyclic nucleotide monophosphate-binding domain, a PDZ (PSD-95/DlgA/ZO-1) domain, a Ras-associating (RA) domain and a Ras exchanger motif. RAPGEF6 is closely related to RAPGEF2 and both proteins exhibit GEF activity specific towards Rap 1 and Rap 2. In addition, RAPGEF6 is capable of binding to M-Ras via its RA domain. Due to alternative splicing events, twoMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité