Anti-CDH12 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-13715R
BOSSBS-13715REA 725 EUR InStock
BOSSBS-13715R
Anti-CDH12 Rabbit Polyclonal Antibody
Anticorps
The cadherins are a family of Ca++-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of tissue structure and morpho-genesis (1-4). Cadherins each contain a large extracellular domain at the amino terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity (5,6). The relatively short carboxy terminal, intracellular domain interacts with a variety of cytoplasmic proteins, including catenin b, to regulate cadherin function (7). BR-cadherin (for brain-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité