Anti-GPR30 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-1380R-CY7
BOSSBS-1380R-CY7EA 909 EUR InStock
BOSSBS-1380R-CY7
Anti-GPR30 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
G-protein coupled estrogen receptor that binds to 17-beta-estradiol (E2) with high affinity, leading to rapid and transient activation of numerous intracellular signaling pathways. Stimulates cAMP production, calcium mobilisation and tyrosine kinase Src inducing the release of heparin-bound epidermal growth factor (HB-EGF) and subsequent transactivation of the epidermal growth factor receptor (EGFR), activating downstream signaling pathways such as PI3K/Akt and ERK/MAPK. Mediates pleiotropic functions among others in the cardiovascular, endocrine, reproductive, immune and central nervousMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité