Anti-CHMP2A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-13913R-A750
BOSSBS-13913R-A750EA 890 EUR InStock
BOSSBS-13913R-A750
Anti-CHMP2A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
Probable core component of the endosomal sorting required for transport complex III (ESCRT-III) which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles (ILVs) that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins, such as stimulated growth factor receptors, lysosomal enzymes and lipids. The MVB pathway appears to require the sequential function of ESCRT-O, -I,-II and -III complexes. ESCRT-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité