Anti-C22orf26 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Fournisseur: Bioss
BS-15134R-A680
BOSSBS-15134R-A680EA 890 EUR InStock
BOSSBS-15134R-A680
Anti-C22orf26 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Chromosome 22 contains over 500 genes and about 49 million bases. Being the second smallest human chromosome, 22 contains a surprising variety of interesting genes. Phelan-McDermid syndrome, neurofibromatosis type 2 and autism are associated with chromosome 22. A schizophrenia susceptibility locus has been identified on chromosome 22 and studies show that 22q11 deletion symptoms include a high incidence of schizophrenia. Translo-acations between chromosomes 9 and 22 may lead to the formation of the Philadelphia chromosome and the subsequent production of the novel fusion protein, Bcr-Abl, aMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité