Anti-C3orf36 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Bioss
BS-15176R-HRP
BOSSBS-15176R-HRPEA 900 EUR InStock
BOSSBS-15176R-HRP
Anti-C3orf36 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
C3orf36 (chromosome 3 open reading frame 36), also known as FLJ22173, MGC125760 or MGC125761, is a 165 amino acid protein encoded by a gene that maps to human chromosome 3p25.3. Chromosome 3 is made up of approximately 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesisMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité