Anti-C6ORF70 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Fournisseur: Bioss
BS-15252R-A555
BOSSBS-15252R-A555EA 893 EUR InStock
BOSSBS-15252R-A555
Anti-C6ORF70 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Anticorps
Making up nearly 6% of the human genome, chromosome 6 contains around 1200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are keyMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité