Anti-ATF4/CREB-2 Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-1531R-CY5.5
BOSSBS-1531R-CY5.5EA 909 EUR InStock
BOSSBS-1531R-CY5.5
Anti-ATF4/CREB-2 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Transcriptional activator. Binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AC][AG]-3'), a sequence present in many viral and cellular promoters. Cooperates with FOXO1 in osteoblasts to regulate glucose homeostasis through suppression of beta-cell production and decrease in insulin production (By similarity). It binds to a Tax-responsive enhancer element in the long terminal repeat of HTLV-I. Regulates the induction of DDIT3/CHOP and asparagine synthetase (ASNS) in response to ER stress. In concert with DDIT3/CHOP, activates the transcription of TRIB3 and promotes ER stress-More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité