Anti-C9ORF30 Rabbit Polyclonal Antibody (FITC)
Fournisseur: Bioss
BS-15326R-FITC
BOSSBS-15326R-FITCEA 900 EUR InStock
BOSSBS-15326R-FITC
Anti-C9ORF30 Rabbit Polyclonal Antibody (FITC)
Anticorps
C9orf30 is a 275 amino acid protein that is expressed in brain and belongs to the UPF0439 family. The gene encoding C9orf30 maps to human chromosome 9q31.1. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 geneMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité