Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Anti-C9ORF84 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Supplier: Bioss
BS-15344R-A680
BOSSBS-15344R-A680EA 890 EUR InStock
BOSSBS-15344R-A680
Anti-C9ORF84 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antibodies
Chromosome 9 consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype. Hereditary hemorrhagic telangiectasia, which is characterised by harmful vascular defects, is associated with the chromosome 9 gene encoding endoglin protein, ENG. Familial dysautonomia is also associated with chromosome 9 though through the gene IKBKAP. Notably, chromosome 9 encompasses theMore Product Information
Order Now
ORDER
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 016 385 011
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 016 385 011.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Customer Service at vwrbe@be.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Products marked with this symbol have been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 016/385 011.
Collapse All
Pack type Pk VWR Catalog Number Quantity
VIA 100 µl BOSSBS-15344R-A680
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory, Enter 'x' if unknown for VWR Chemicals
SPECIFICATIONS
Documentation
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty