Anti-GPCR RDC1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Fournisseur: Bioss
BS-15371R-A488
BOSSBS-15371R-A488EA 893 EUR InStock
BOSSBS-15371R-A488
Anti-GPCR RDC1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Anticorps
Atypical chemokine receptor that controls chemokine levels and localisation via high-affinity chemokine binding that is uncoupled from classic ligand-driven signal transduction cascades, resulting instead in chemokine sequestration, degradation, or transcytosis. Also known as interceptor (internalising receptor) or chemokine-scavenging receptor or chemokine decoy receptor. Acts as a receptor for chemokines CXCL11 and CXCL12/SDF1. Chemokine binding does not activate G-protein-mediated signal transduction but instead induces beta-arrestin recruitment, leading to ligand internalisation andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité