Anti-CHIP Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Fournisseur: Bioss
BS-15405R-A647
BOSSBS-15405R-A647EA 893 EUR InStock
BOSSBS-15405R-A647
Anti-CHIP Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Anticorps
E3 ubiquitin-protein ligase which targets misfolded chaperone substrates towards proteasomal degradation. Collaborates with ATXN3 in the degradation of misfolded chaperone substrates: ATXN3 restricting the length of ubiquitin chain attached to STUB1/CHIP substrates and preventing further chain extension. Ubiquitinates NOS1 in concert with Hsp70 and Hsp40. Modulates the activity of several chaperone complexes, including Hsp70, Hsc70 and Hsp90. Mediates transfer of non-canonical short ubiquitin chains to HSPA8 that have no effect on HSPA8 degradation. Mediates polyubiquitination of DNAMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité