Anti-HDPTP Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Fournisseur: Bioss
BS-15434R-A488
BOSSBS-15434R-A488EA 893 EUR InStock
BOSSBS-15434R-A488
Anti-HDPTP Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Anticorps
HD-PTP is a 1636 amino acid protein encoded by the human gene PTPN23. HD-PTP belongs to the protein-tyrosine phosphatase family, non-receptor class subfamily. It contains one BRO1 domain, two TPR repeats and one tyrosine-protein phosphatase domain. The C-terminal region contains the PTP-like domain, whereas the N-terminal region contains the two TPR regions. These regions are homologous to the yeast protein, BRO1, which is involved in the mitogen-activated protein kinase signaling pathway. Similarly, HD-PTP is believed to act as a negative regulator of Ras-mediated mitogenic activity and isMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité