Anti-Shh Rabbit Polyclonal Antibody (Cy3®)
Fournisseur: Bioss
BS-1544R-CY3
BOSSBS-1544R-CY3EA 909 EUR InStock
BOSSBS-1544R-CY3
Anti-Shh Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Intercellular signal essential for a variety of patterning events during development: signal produced by the notochord that induces ventral cell fate in the neural tube and somites, and the polarizing signal for patterning of the anterior-posterior axis of the developing limb bud. Displays both floor plate- and motor neuron-inducing activity. The threshold concentration of N-product required for motor neuron induction is 5-fold lower than that required for floor plate induction. Activates the transcription of target genes by interacting with its receptor PTCH1 to prevent normal inhibition byMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité