Anti-Syncytin 2 Rabbit Polyclonal Antibody (HRP)
Fournisseur: Bioss
BS-15466R-HRP
BOSSBS-15466R-HRPEA 883 EUR InStock
BOSSBS-15466R-HRP
Anti-Syncytin 2 Rabbit Polyclonal Antibody (HRP)
Anticorps
This endogenous retroviral envelope protein has retained its original fusogenic properties and participates in trophoblast fusion and the formation of a syncytium during placenta morphogenesis. The interaction with MFSD2A is apparently important for this process. Endogenous envelope proteins may have kept, lost or modified their original function during evolution but this one can still make pseudotypes with MLV, HIV-1 or SIV-1 virions and confer infectivity. Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. The surface protein mediatesMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité