Anti-HEXB chain B Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Fournisseur: Bioss
BS-15468R-A555
BOSSBS-15468R-A555EA 893 EUR InStock
BOSSBS-15468R-A555
Anti-HEXB chain B Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Anticorps
Hexosaminidase B is the beta subunit of the lysosomal enzyme beta-hexosaminidase that, together with the cofactor GM2 activator protein, catalyses the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Beta-hexosaminidase is composed of two subunits, alpha and beta, which are encoded by separate genes. Both beta-hexosaminidase alpha and beta subunits are members of family 20 of glycosyl hydrolases. Mutations in the alpha or beta subunit genes lead to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité