Anti-T122 Rabbit Polyclonal Antibody (HRP)
Fournisseur: Bioss
BS-15558R-HRP
BOSSBS-15558R-HRPEA 900 EUR InStock
BOSSBS-15558R-HRP
Anti-T122 Rabbit Polyclonal Antibody (HRP)
Anticorps
IFT122 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptionalMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité