Anti-IGLK Rabbit Polyclonal Antibody (FITC)
Fournisseur: Bioss
BS-15576R-FITC
BOSSBS-15576R-FITCEA 900 EUR InStock
BOSSBS-15576R-FITC
Anti-IGLK Rabbit Polyclonal Antibody (FITC)
Anticorps
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. Alternative splicing of this gene results in three tissue-specific forms of glucokinase, one found in pancreatic islet beta cells and two found in liver. The protein localises to the outer membrane of mitochondria. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. Mutations in this gene have been associated with non-insulin dependent diabetes mellitus (NIDDM), maturity onsetMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité