Anti-TEM8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Fournisseur: Bioss
BS-15583R-A750
BOSSBS-15583R-A750EA 890 EUR InStock
BOSSBS-15583R-A750
Anti-TEM8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
This gene encodes a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colourectal cancer. The encoded protein has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax. The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to this receptor protein mediates delivery of toxin components to the cytosol of cells. Once inside the cell, the other two components of anthrax toxin, edoema factor (EF) and lethal factor (LF) disrupt normal cellularMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité