Anti-AVP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Fournisseur: Bioss
BS-1930R-A488
BOSSBS-1930R-A488EA 876 EUR InStock
BOSSBS-1930R-A488
Anti-AVP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorps
Vasopressin, also known as arginine vasopressin (AVP) or antidiuretic hormone (ADH), is a posterior pituitary hormone that is synthesised in the hypothalamus. Vasopressin is synthesised as a precursor protein that consists of arginine vasopressin and two associated proteins, neurophysin 2 and the glycopeptide copeptin. Vasopressin, together with its carrier protein neurophysin II, is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. Vasopressin acts as a growth factor byMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité