Anti-EGF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-2009R-A350
BOSSBS-2009R-A350EA 858 EUR InStock
BOSSBS-2009R-A350
Anti-EGF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture. Magnesiotropic hormone that stimulates magnesium reabsorption in the renal distal convoluted tubule via engagement of EGFR and activation of the magnesium channel TRPM6 (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité