Anti-S100 B Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-2015R-CY7
BOSSBS-2015R-CY7EA 909 EUR InStock
BOSSBS-2015R-CY7
Anti-S100 B Rabbit Polyclonal Antibody (Cy7®)
Anticorps
Weakly binds calcium but binds zinc very tightly-distinct binding sites with different affinities exist for both ions on each monomer. Physiological concentrations of potassium ion antagonize the binding of both divalent cations, especially affecting high-affinity calcium-binding sites. Binds to and initiates the activation of STK38 by releasing autoinhibitory intramolecular interactions within the kinase. Interaction with AGER after myocardial infarction may play a role in myocyte apoptosis by activating ERK1/2 and p53/TP53 signaling. Could assist ATAD3A cytoplasmic processing, preventingMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité