Anti-PAFAH1B1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-2035R-A555
BOSSBS-2035R-A555EA 893 EUR InStock
BOSSBS-2035R-A555
Anti-PAFAH1B1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
Required for proper activation of Rho GTPases and actin polymerization at the leading edge of locomoting cerebellar neurons and postmigratory hippocampal neurons in response to calcium influx triggered via NMDA receptors. Non-catalytic subunit of an acetylhydrolase complex which inactivates platelet-activating factor (PAF) by removing the acetyl group at the SN-2 position (By similarity). Positively regulates the activity of the minus-end directed microtubule motor protein dynein. May enhance dynein-mediated microtubule sliding by targeting dynein to the microtubule plus end. Required forMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité