Anti-CALD1 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-2082R-CY5
BOSSBS-2082R-CY5EA 905 EUR InStock
BOSSBS-2082R-CY5
Anti-CALD1 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
This gene encodes a calmodulin- and actin-binding protein that plays an essential role in the regulation of smooth muscle and nonmuscle contraction. The conserved domain of this protein possesses the binding activities to Ca(2+)-calmodulin, actin, tropomyosin, myosin, and phospholipids. This protein is a potent inhibitor of the actin-tropomyosin activated myosin MgATPase, and serves as a mediating factor for Ca(2+)-dependent inhibition of smooth muscle contraction. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Jul 200More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité