Anti-RNF126 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-2125R-CY5
BOSSBS-2125R-CY5EA 900 EUR InStock
BOSSBS-2125R-CY5
Anti-RNF126 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
E3 ubiquitin-protein ligase that regulates several biological processes through ubiquitination of various target proteins. Depending on the associated E2 ligase, mediates 'Lys-48'- and 'Lys-63'-linked polyubiquitination of substrates. Through their polyubiquitination, may play a role in the endosomal sorting and degradation of several membrane receptors including EGFR, FLT3, MET and CXCR4. May also be part of a BAG6-dependent quality control process ensuring that proteins of the secretory pathway that are mislocalized to the cytosol are degraded by the proteasome. May provide the ubiquitinMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité