Anti-ADAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-2167R-A555
BOSSBS-2167R-A555EA 893 EUR InStock
BOSSBS-2167R-A555
Anti-ADAR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
Catalyzes the hydrolytic deamination of adenosine to inosine in double-stranded RNA (dsRNA) referred to as A-to-I RNA editing. This may affect gene expression and function in a number of ways that include mRNA translation by changing codons and hence the amino acid sequence of proteins; pre-mRNA splicing by altering splice site recognition sequences; RNA stability by changing sequences involved in nuclease recognition; genetic stability in the case of RNA virus genomes by changing sequences during viral RNA replication; and RNA structure-dependent activities such as microRNA production orMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité