Anti-GZMA Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-2578R-CY5.5
BOSSBS-2578R-CY5.5EA 900 EUR InStock
BOSSBS-2578R-CY5.5
Anti-GZMA Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Abundant protease in the cytosolic granules of cytotoxic T-cells and NK-cells which activates caspase-independent cell death with morphological features of apoptosis when delivered into the target cell through the immunological synapse. It cleaves after Lys or Arg. Cleaves APEX1 after 'Lys-31' and destroys its oxidative repair activity. Cleaves the nucleosome assembly protein SET after 'Lys-189', which disrupts its nucleosome assembly activity and allows the SET complex to translocate into the nucleus to nick and degrade the DNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité