Anti-AURKA Rabbit Polyclonal Antibody (Cy5.5®)
Fournisseur: Bioss
BS-2751R-CY5.5
BOSSBS-2751R-CY5.5EA 900 EUR InStock
BOSSBS-2751R-CY5.5
Anti-AURKA Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment.Aurora B is a mitotic protein kinase that phosphorylates histone H3 (probably on Serine 10), behaves as a chromosomal passenger protein, and may regulate several stages of mitosis such asMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité