Anti-CRK Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-2763R-CY5
BOSSBS-2763R-CY5EA 900 EUR InStock
BOSSBS-2763R-CY5
Anti-CRK Rabbit Polyclonal Antibody (Cy5®)
Anticorps
Crk p38 (CrkII) is an isoform of the protein Crk. CrkI and CrkII are produced from the same crk gene by alternative splicing. The two isoforms differ in their biological activities with CrkII having less transforming activity than CrkI.Crk p38 is a member of an adapter protein family that binds to several tyrosine-phosphorylated proteins. It has several SH2 and SH3 domains (src-homology domains) and is involved in several signaling pathways, recruiting cytoplasmic proteins in the vicinity of tyrosine kinase through SH2-phosphotyrosine interaction. The N-terminal SH2 domain of this proteinMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité