Anti-HDAC4 Rabbit Polyclonal Antibody
Fournisseur: Bioss
BS-2809R
BOSSBS-2809REA 725 EUR InStock
BOSSBS-2809R
Anti-HDAC4 Rabbit Polyclonal Antibody
Anticorps
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. Involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer.
WB(1:100-1000), IHC-P(1:100More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité