Anti-ASB2 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-2835R-CY5
BOSSBS-2835R-CY5EA 900 EUR InStock
BOSSBS-2835R-CY5
Anti-ASB2 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
The ankyrin repeat and SOCS box containing (ASB) protein family is comprised of members which each contain "SOCS boxes" but differ from other families of protein in the motifs they contain upstream of the SOCS box. SOCS boxes are carboxy terminal regions of homology found in the suppressor of cytokine signaling family of proteins. The box region is thought to be the point of interaction between SOCS proteins and E3 ubiquitin ligases. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targetingMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité