Anti-APP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-3028R-A350
BOSSBS-3028R-A350EA 921 EUR InStock
BOSSBS-3028R-A350
Anti-APP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
This gene encodes a cell surface receptor and transmembrane precursor protein that is cleaved by secretases to form a number of peptides. Some of these peptides are secreted and can bind to the acetyltransferase complex APBB1/TIP60 to promote transcriptional activation, while others form the protein basis of the amyloid plaques found in the brains of patients with Alzheimer disease. Mutations in this gene have been implicated in autosomal dominant Alzheimer disease and cerebroarterial amyloidosis (cerebral amyloid angiopathy). Multiple transcript variants encoding several different isoformsMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité